Skip to Content
Merck
All Photos(1)

Key Documents

ENA321765936

Enamine

2-acetamido-6-fluorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
197.17
MDL number:

mp

199-200

Iupac Name

2-(acetylamino)-6-fluorobenzoic acid

Inchi Code

1S/C9H8FNO3/c1-5(12)11-7-4-2-3-6(10)8(7)9(13)14/h2-4H,1H3,(H,11,12)(H,13,14)

InChI key

IWTHKNXLQDASPO-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-77811

Melting Point

199-200

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-77811


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service