Skip to Content
Merck

COMH303775CF

Combi-Blocks, Inc.

2-Ethoxy-1-nitro-3-(trifluoromethoxy)benzene

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
EUR 540.00

EUR 540.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
EUR 540.00

About This Item

CAS Number:
Molecular Weight:
251.16
MDL number:

EUR 540.00


Please contact Customer Service for Availability

Iupac Name

2-ethoxy-1-nitro-3-(trifluoromethoxy)benzene

Inchi Code

1S/C9H8F3NO4/c1-2-16-8-6(13(14)15)4-3-5-7(8)17-9(10,11)12/h3-5H,2H2,1H3

InChI key

NOKQCFPFIBBDIH-UHFFFAOYSA-N

Purity

96%

Country of Origin

CN

Product Link

https://www.combi-blocks.com/cgi-bin/find.cgi?HC-4519

MSDS

https://www.combi-blocks.com/cgi-bin/find.cgi?HC-4519


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service