Skip to Content
Merck
All Photos(1)

Key Documents

CIAH9884F755

ChemScene LLC (Preferred Partner)

Thalidomide-O-amido-PEG2-C2-NH2 (hydrochloride)

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$399.30
50 MG
$537.30
100 MG
$735.68
250 MG
$1,513.65

Vendor SKU: CS-0120478-25MG

$399.30


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
25 MG
$399.30
50 MG
$537.30
100 MG
$735.68
250 MG
$1,513.65

About This Item

CAS Number:
Molecular Weight:
498.92
MDL number:

Vendor SKU: CS-0120478-25MG

$399.30


Ships in 14 days - from Aldrich Partner.

Iupac Name

N-(2-(2-(2-aminoethoxy)ethoxy)ethyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)oxy)acetamide hydrochloride

Inchi Code

1S/C21H26N4O8.ClH/c22-6-8-31-10-11-32-9-7-23-17(27)12-33-15-3-1-2-13-18(15)21(30)25(20(13)29)14-4-5-16(26)24-19(14)28;/h1-3,14H,4-12,22H2,(H,23,27)(H,24,26,28);1H

InChI key

CPJQBYYTYFGXKD-UHFFFAOYSA-N

storage temp.

4C

Salt Data

Hydrochloride

Purity

98%

Country of Origin

CN

Physical Form

Solid

Product Link

https://www.chemscene.com/2376990-30-4.html

MSDS

https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-0120478.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service