Skip to Content
Merck
All Photos(1)

Key Documents

C12H95E92550

Chemcia Scientific, LLC

(2-Chloro-6-trifluoromethyl-pyridin-3-yl)-dimethyl-amine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$623.55
1 G
$892.65

Vendor SKU: BB20-1378-500MG

$623.55


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
500 MG
$623.55
1 G
$892.65

About This Item

Molecular Weight:
224.61

Vendor SKU: BB20-1378-500MG

$623.55


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-chloro-N,N-dimethyl-6-(trifluoromethyl)pyridin-3-amine

Inchi Code

1S/C8H8ClF3N2/c1-14(2)5-3-4-6(8(10,11)12)13-7(5)9/h3-4H,1-2H3

InChI key

SWEDEAJBJBZUBJ-UHFFFAOYSA-N

storage temp.

0-5C

Purity

97%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Light Yellow Liquid

Product Link

http://www.chemcia.com/product_fl02.asp?id=17092

MSDS

http://www.chemcia.com/product_fl02.asp?id=17092


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service