Skip to Content
Merck
All Photos(1)

Key Documents

C12H774CD835

Chemcia Scientific, LLC

7-Bromo-8-methyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$861.60
1 G
$1,720.65
2.5 G
$3,490.50

Vendor SKU: BB20-8247-250MG

$861.60


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$861.60
1 G
$1,720.65
2.5 G
$3,490.50

About This Item

CAS Number:
Molecular Weight:
229.08

Vendor SKU: BB20-8247-250MG

$861.60


Ships in 14 days - from Aldrich Partner.

Iupac Name

7-bromo-8-methyl-2,3-dihydro-1H-pyrido[2,3-b][1,4]oxazine

Inchi Code

1S/C8H9BrN2O/c1-5-6(9)4-11-8-7(5)10-2-3-12-8/h4,10H,2-3H2,1H3

InChI key

ZHHLHPRVXSRETL-UHFFFAOYSA-N

storage temp.

0-5C

Purity

95%

Country of Origin

US

Shipping Temperature

Ambient

Physical Form

Light Yellow Solid

Product Link

http://www.chemcia.com/product_fl02.asp?id=13968

MSDS

http://www.chemcia.com/product_fl02.asp?id=13968


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service