Skip to Content
Merck
All Photos(1)

Key Documents

CH4H8C2C85AF

ChemBridge Corporation

methyl (3-bromo-5-hydroxy-1H-1,2,4-triazol-1-yl)acetate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$265.44
1 G
$393.78
5 G
$1,350.12

Vendor SKU: 4110510-250MG

$265.44


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$265.44
1 G
$393.78
5 G
$1,350.12

About This Item

CAS Number:
Molecular Weight:
236.02
MDL number:

Vendor SKU: 4110510-250MG

$265.44


Ships in 14 days - from Aldrich Partner.

Iupac Name

methyl 2-(3-bromo-5-hydroxy-1H-1,2,4-triazol-1-yl)acetate

Inchi Code

1S/C5H6BrN3O3/c1-12-3(10)2-9-5(11)7-4(6)8-9/h2H2,1H3,(H,7,8,11)

InChI key

YYNNAFSZZVLDCB-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

solid

Product Link

https://www.hit2lead.com/building-blocks/4110510

MSDS

https://www.hit2lead.com/sds/4110510_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service