Skip to Content
Merck
All Photos(1)

Key Documents

CH4454669633

ChemBridge Corporation

tert-butyl rac-(3S,4R)-3-amino-4-hydroxy-1-pyrrolidinecarboxylate hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$462.09
1 G
$758.10
5 G
$2,807.40
10 G
$4,515.15

Vendor SKU: 4072089-250MG

$462.09


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$462.09
1 G
$758.10
5 G
$2,807.40
10 G
$4,515.15

About This Item

Molecular Weight:
238.71
MDL number:

Vendor SKU: 4072089-250MG

$462.09


Ships in 14 days - from Aldrich Partner.

Iupac Name

tert-butyl (3S,4R)-3-amino-4-hydroxypyrrolidine-1-carboxylate hydrochloride

Inchi Code

1S/C9H18N2O3.ClH/c1-9(2,3)14-8(13)11-4-6(10)7(12)5-11;/h6-7,12H,4-5,10H2,1-3H3;1H/t6-,7+;/m0./s1

InChI key

RTJTVYMOCPYEAG-UOERWJHTSA-N

storage temp.

RT

Salt Data

HCl

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

solid

Product Link

https://www.hit2lead.com/building-blocks/4072089

MSDS

https://www.hit2lead.com/sds/4072089_sds.pdf


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service