Skip to Content
Merck
All Photos(1)

Documents

D-140

Supelco

(±)-threo-3,4-Dichloromethylphenidate HCl

1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17Cl2NO2 · HCl
CAS Number:
Molecular Weight:
338.66
UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

format

single component solution

storage temp.

−20°C

SMILES string

O=C(OC)[C@@H]([C@@]1([H])CCCCN1)C2=CC=C(Cl)C(Cl)=C2.[H]Cl

InChI

1S/C14H17Cl2NO2.ClH/c1-19-14(18)13(12-4-2-3-7-17-12)9-5-6-10(15)11(16)8-9;/h5-6,8,12-13,17H,2-4,7H2,1H3;1H/t12-,13-;/m1./s1

InChI key

DJQISAOWJXQDPU-OJERSXHUSA-N

General description

3,4-Dichloromethylphenidate, also known as 3,4-CTMP, is a structural analogue of methylphenidate (Ritalin). 3,4-CTMP is reportedly seven times more potent than methylphenidate and is sold online as a novel psychoactive stimulant. This certified Snap-N-Spike® solution standard is applicable for use in LC/MS or GC/MS applications such as urine drug testing, clinical toxicology and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service