Skip to Content
Merck
All Photos(1)

Key Documents

A-181

Supelco

Glutaryl-L-Carnitine-(N-Methyl-D3) lithium salt solution

100 μg/mL (90:10 Methanol: 1% aqueous formic acid), certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12D3H17NO6 · Li
CAS Number:
Molecular Weight:
284.25
MDL number:
UNSPSC Code:
41116107
Product A-181 is not currently sold in your country. Contact Technical Service

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL (90:10 Methanol: 1% aqueous formic acid)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

SMILES string

C[N+](C)(C([2H])([2H])[2H])C[C@H](OC(CCCC([O-])=O)=O)CC([O-])=O.[Li+]

General description

A stable-labeled internal standard of routinely tested Glutaryl-L-Carnitine at a concentration of 100 μg/mL. Acylcarnitines are biomarker for many inborn errors of metabolism that cause several metabolic diseases and are routinely tested in newborn screening programs. This stable-labeled internal standard is suitable for critical quantitative testing applications by LC/MS or GC/MS in research and clinical environments. The stable, quantitative solution format provides significant benefits to the user such as convenience of use, material savings, and increased efficiency of labor. Certified concentration value provides increased efficiency from less time remaking pools due to dilution errors or rerunning samples.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

In this article we provide a complete workflow of quantification and identification of acylcarnitines in dried blood spots (DBS) using stable isotope-labeled (SIL) acylcarnitines CRMs by FIA-MS/MS.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service