Skip to Content
Merck
All Photos(2)

Key Documents

810267P

Avanti

C11 TopFluor Glucosyl Ceramide

Avanti Research - A Croda Brand 810267P, powder

Synonym(s):

N-[11-(dipyrrometheneboron difluoride)undecanoyl]-D-glucosyl-β1-1′-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing

Select a Size

250 μG
$782.00

$782.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
250 μG
$782.00

About This Item

Empirical Formula (Hill Notation):
C48H80N3O8F2
CAS Number:
Molecular Weight:
865.16
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$782.00


Estimated to ship on26 April 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 250 μg (810267P-250ug)

manufacturer/tradename

Avanti Research - A Croda Brand 810267P

shipped in

dry ice

storage temp.

−20°C

General description

Glucosyl Ceramide (GlcCer)/cerebroside/galactosylceramide is a neutral, polar lipid. It can be ionized delicately in negative-and positive-ion modes.[1] It is the simplest glycosphingolipids (GSLs), that is present abundantly in the mammalian tissues. GlcCer is produced through glycosylation of Cer with the help of GlcCer synthase (GCS) at the cytosolic leaflet of Golgi apparatus.[2]

Application

C11 TopFluor Glucosyl Ceramide is suitable to use as a label in the GLM II model and as a component in the usual giant unilamellar vesicle (GUV) lipid mixture (GLM).[3]

Biochem/physiol Actions

Glucosyl ceramide (GlcCer) plays a major role in the pathways of glycosphingolipid metabolism. It helps to modulate cell physiology. It acts as an intracellular messenger in cell maintenance and regulation.[2]

Packaging

5 mL Amber Glass Screw Cap Vial (810267P-250ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC
TopFluor is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lipid segregation and membrane budding induced by the peripheral membrane binding protein annexin A2
Drucker P, et al.
Test, 288(34), 24764-24776 (2013)
Characterization of Lipids by Electrospray Ionization Mass Spectrometry, Lipid Analysis
Lipid analysis (2012)
Effect of glucosylceramide on the biophysical properties of fluid membranes
Varela ARP, et al.
Biochimica et Biophysica Acta - Biomembranes, 1828(3), 1122-1130 (2013)
Omid Motabar et al.
Analytical and bioanalytical chemistry, 402(2), 731-739 (2011-10-29)
Glucocerebrosidase is a lysosomal enzyme that catalyzes the hydrolysis of glucosylceramide to form ceramide and glucose. A deficiency of lysosomal glucocerebrosidase due to genetic mutations results in Gaucher disease, in which glucosylceramide accumulates in the lysosomes of certain cell types.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service