Skip to Content
Merck
All Photos(2)

Key Documents

700133P

Avanti

3β,25-OH-7-oxo-5-cholestenoic acid

Avanti Research - A Croda Brand 700133P, powder

Synonym(s):

3β,25-dihydroxy-7-oxo-5-cholestenoic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
$1,450.00

$1,450.00


Estimated to ship on27 April 2025


Request a Bulk Order

Select a Size

Change View
1 MG
$1,450.00

About This Item

Empirical Formula (Hill Notation):
C27H42 O5
CAS Number:
Molecular Weight:
446.62
UNSPSC Code:
12352211
NACRES:
NA.25

$1,450.00


Estimated to ship on27 April 2025


Request a Bulk Order

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (700133P-1mg)

manufacturer/tradename

Avanti Research - A Croda Brand 700133P

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H](C1)CC[C@](C1=C[C@H]2O)(C)C3C2C(CC[C@@H]4[C@H](C)CCCC(C)(O)CO)[C@]4(C)CC3

InChI key

JGEBMCVWDISKRI-MRAVHIDRSA-N

General description

3β,25-OH-7-oxo-5-cholestenoic acid is a cholesterol derivative and an oxysterol derived from cholesterol. The synthesis of CA occurs in mitochondria and the enzyme sterol 27-hydroxylase (CYP27A1) catalyzes the step.[1]

Biochem/physiol Actions

Cholestenoic acid (CA) serves as a ligand to nuclear receptor and may be neurotoxic or neuroprotective based on their structural features.[2] Defects in the enzymes catalyzing the formation of cholestenoic acids are implicated in disorders related to motor neuron degeneration.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (700133P-1mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spyridon Theofilopoulos et al.
The Journal of clinical investigation, 124(11), 4829-4842 (2014-10-02)
Cholestenoic acids are formed as intermediates in metabolism of cholesterol to bile acids, and the biosynthetic enzymes that generate cholestenoic acids are expressed in the mammalian CNS. Here, we evaluated the cholestenoic acid profile of mammalian cerebrospinal fluid (CSF) and
Cholestenoic acid is a prognostic biomarker in acute respiratory distress syndrome.
Jennifer H Madenspacher et al.
The Journal of allergy and clinical immunology, 143(1), 440-442 (2018-10-09)
Peter J Crick et al.
The Journal of steroid biochemistry and molecular biology, 195, 105475-105475 (2019-09-22)
While the presence and abundance of the major oxysterols and cholestenoic acids in the circulation is well established, minor cholesterol metabolites may also have biological importance and be of value to investigate. In this study by observing the metabolism of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service