ATEH961DE235
AstaTech, Inc.
(2-HYDROXYQUINOLIN-3-YL)BORONIC ACID
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-oxo-1,2-dihydro-3-quinolinylboronic acid
Inchi Code
1S/C9H8BNO3/c12-9-7(10(13)14)5-6-3-1-2-4-8(6)11-9/h1-5,13-14H,(H,11,12)
InChI key
RIRFRJDLNNSOJT-UHFFFAOYSA-N
Purity
95%
Country of Origin
CN
Physical Form
Pale-yellow to Yellow-brown Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=127742
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=W17048
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service