Skip to Content
Merck
All Photos(1)

Key Documents

ATEH03AB4FDA

AstaTech, Inc.

1-(1-(3-BROMOPHENYL)ETHYL)PIPERIDIN-4-OL

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$506.25
250 MG
$1,366.59
1 G
$3,725.53

Vendor SKU: N11660-50MG

$506.25


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
$506.25
250 MG
$1,366.59
1 G
$3,725.53

About This Item

CAS Number:
Molecular Weight:
284.2
MDL number:

Vendor SKU: N11660-50MG

$506.25


Ships in 14 days - from Aldrich Partner.

Iupac Name

1-(1-(3-bromophenyl)ethyl)piperidin-4-ol

Inchi Code

1S/C13H18BrNO/c1-10(11-3-2-4-12(14)9-11)15-7-5-13(16)6-8-15/h2-4,9-10,13,16H,5-8H2,1H3

InChI key

KLVAXUDXTLUENV-UHFFFAOYSA-N

Purity

95%

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=115730

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=N11660


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service