Skip to Content
Merck
All Photos(1)

Key Documents

ATEH03AB3EDF

AstaTech, Inc.

DIETHYL 2-(2,2,2-TRIFLUOROACETYL)SUCCINATE

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
270.21
MDL number:

Iupac Name

diethyl 2-(2,2,2-trifluoroacetyl)succinate

Inchi Code

1S/C10H13F3O5/c1-3-17-7(14)5-6(9(16)18-4-2)8(15)10(11,12)13/h6H,3-5H2,1-2H3

InChI key

VFUJVMLDXHUHOP-UHFFFAOYSA-N

Purity

95%

Physical Form

Pale-yellow to Yellow-brown Liquid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=114555

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=S11596


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service