Skip to Content
Merck
All Photos(1)

Key Documents

ATE517251966

AstaTech, Inc.

3-[(1R)-3-[BIS(1-METHYLETHYL)AMINO]-1-PHENYLPROPYL]-4-HYDROXYBENZENEMETHANOL

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$391.97
25 MG
$780.09
100 MG
$2,224.78

Vendor SKU: F31141-5MG

$391.97


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
5 MG
$391.97
25 MG
$780.09
100 MG
$2,224.78

About This Item

CAS Number:
Molecular Weight:
341.49
MDL number:

Vendor SKU: F31141-5MG

$391.97


Ships in 14 days - from Aldrich Partner.

Iupac Name

2-[(1R)-3-(diisopropylamino)-1-phenylpropyl]-4-(hydroxymethyl)phenol

Inchi Code

1S/C22H31NO2/c1-16(2)23(17(3)4)13-12-20(19-8-6-5-7-9-19)21-14-18(15-24)10-11-22(21)25/h5-11,14,16-17,20,24-25H,12-13,15H2,1-4H3/t20-/m1/s1

InChI key

DUXZAXCGJSBGDW-HXUWFJFHSA-N

Purity

95%

Physical Form

Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=92621

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=F31141


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service