Skip to Content
Merck
All Photos(1)

Key Documents

ATE361619674

AstaTech, Inc.

(S)-N1,N1-DIMETHYL-3-PHENYLPROPANE-1,2-DIAMINE 2HCL

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
251.2
MDL number:

Iupac Name

(2S)-N~1~,N~1~-dimethyl-3-phenyl-1,2-propanediamine dihydrochloride

Inchi Code

1S/C11H18N2.2ClH/c1-13(2)9-11(12)8-10-6-4-3-5-7-10;;/h3-7,11H,8-9,12H2,1-2H3;2*1H/t11-;;/m0../s1

InChI key

ZURBYNBDHKDKQY-IDMXKUIJSA-N

storage temp.

2-8 C

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

Pale-yellow to Yellow-brown Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=3633

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=53132


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service