ATE341033502
AstaTech, Inc.
4-(3,4-DICHLOROPHENOXY)BENZENESULFONYL CHLORIDE
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
mp
86-88 C
Iupac Name
4-(3,4-dichlorophenoxy)benzenesulfonyl chloride
Inchi Code
1S/C12H7Cl3O3S/c13-11-6-3-9(7-12(11)14)18-8-1-4-10(5-2-8)19(15,16)17/h1-7H
InChI key
WSEZTLINFKIBOX-UHFFFAOYSA-N
storage temp.
2-8 C
Purity
95%
Shipping Temperature
Normal
Physical Form
White to Yellow Solid
Product Link
http://astatechinc.com/CPSResult.php?CRNO=10153
Melting Point
86-88 C
Signal Word
Danger
Hazard Statements
Precautionary statements
MSDS
http://astatechinc.com/Asta_MSDS.php?Catalog=AR1964
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service