Skip to Content
Merck
All Photos(1)

Key Documents

APOH30347AD8

Apollo Scientific Ltd

4-Bromo-2-chloro-5-iodophenol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
333.35
MDL number:

Pricing and availability is not currently available. Contact Technical Service

Iupac Name

4-bromo-2-chloro-5-iodophenol

Inchi Code

1S/C6H3BrClIO/c7-3-1-4(8)6(10)2-5(3)9/h1-2,10H

InChI key

CRTGJCONBCCZTQ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

95%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/4-bromo-2-chloro-5-iodophenol

Pictograms

Signal Word

Danger

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-bromo-2-chloro-5-iodophenol


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service