Skip to Content
Merck
All Photos(1)

Key Documents

APO455831013

Apollo Scientific Ltd

3-Iodobenzene-1,2-diamine

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$328.95
1 G
$628.50

Vendor SKU: OR43514-250MG

$328.95


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
250 MG
$328.95
1 G
$628.50

About This Item

CAS Number:
Molecular Weight:
234.04
MDL number:

Vendor SKU: OR43514-250MG

$328.95


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

bp

49-51

Iupac Name

3-iodo-1,2-benzenediamine

Inchi Code

1S/C6H7IN2/c7-4-2-1-3-5(8)6(4)9/h1-3H,8-9H2

InChI key

ZPGYMGXNLJHVHS-UHFFFAOYSA-N

storage temp.

Store at 2-8C

Purity

97%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/3-iodobenzene-12-diamine

Flash Point

144/5mm

Boiling point

49-51

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/3-iodobenzene-12-diamine


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service