APO455829201
Apollo Scientific Ltd
Diethyl 2-cyano-3-oxosuccinate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
88-90
Iupac Name
diethyl 2-cyano-3-oxosuccinate
Inchi Code
1S/C9H11NO5/c1-3-14-8(12)6(5-10)7(11)9(13)15-4-2/h6H,3-4H2,1-2H3
InChI key
FTSVKFGREWPODP-UHFFFAOYSA-N
storage temp.
Ambient Temperature
Country of Origin
GB
Shipping Temperature
Ambient Temperature
Physical Form
Solid
Product Link
https://store.apolloscientific.co.uk/product/diethyl-2-cyano-3-oxosuccinate
Boiling point
88-90
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://store.apolloscientific.co.uk/product/diethyl-2-cyano-3-oxosuccinate
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service