Skip to Content
Merck
All Photos(1)

Key Documents

APO423213883

Apollo Scientific Ltd

6-Fluoro-2-hydroxy-3-nitrobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
201.11
MDL number:

Iupac Name

6-fluoro-2-hydroxy-3-nitrobenzoic acid

Inchi Code

1S/C7H4FNO5/c8-3-1-2-4(9(13)14)6(10)5(3)7(11)12/h1-2,10H,(H,11,12)

InChI key

UERRYAMICLGJBJ-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Solid

Product Link

https://store.apolloscientific.co.uk/product/6-fluoro-2-hydroxy-3-nitrobenzoic-acid

Synonym

6-Fluoro-3-nitrosalicylic acid, 2-Carboxy-3-fluoro-6-nitrophenol

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/6-fluoro-2-hydroxy-3-nitrobenzoic-acid


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service