Skip to Content
Merck

AMBH98C8DC33

Ambeed, Inc.

6-Bromo-4-methylnicotinic acid

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
216.03
MDL number:

Pricing and availability is not currently available.

Iupac Name

6-bromo-4-methylnicotinic acid

Inchi Code

1S/C7H6BrNO2/c1-4-2-6(8)9-3-5(4)7(10)11/h2-3H,1H3,(H,10,11)

InChI key

MOCUEPIHCCYSMT-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
W291102P49104P49805
Piperonal 99%

P49104

Piperonal

Quality Level

100

Quality Level

400

Quality Level

100

Quality Level

100

form

solid

form

-

form

solid

form

powder

mp

50-54 °C (lit.)

mp

35-39 °C (lit.)

mp

35-39 °C (lit.)

mp

229-231 °C (lit.)

storage temp.

Inert atmosphere,2-8C

Purity

97%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

solid

Product Link

https://www.ambeed.com/products/1060804-74-1.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.ambeed.com/products/1060804-74-1.html


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service