Skip to Content
Merck
All Photos(1)

Key Documents

AMBH93EDD2C9

Ambeed, Inc.

1-Chloro-4-methoxybutane

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$94.84
25 G
$98.81
100 G
$118.64

Vendor SKU: A767437-10G

$94.84


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
10 G
$94.84
25 G
$98.81
100 G
$118.64

About This Item

CAS Number:
Molecular Weight:
122.59
MDL number:

Vendor SKU: A767437-10G

$94.84


Ships in 14 days - from Aldrich Partner.

Notify Me

Get notified when this item is ready to ship via email.

Iupac Name

1-chloro-4-methoxybutane

Inchi Code

1S/C5H11ClO/c1-7-5-3-2-4-6/h2-5H2,1H3

InChI key

DFLRARJQZRCCKN-UHFFFAOYSA-N

storage temp.

Sealed in dry,Room Temperature

Purity

98%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.ambeed.com/products/17913-18-7.html

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary statements


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service