Skip to Content
Merck
All Photos(1)

Key Documents

AMBH6FD7F7DA

Ambeed, Inc.

tert-Butyl 2-ethylhydrazinecarboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
$237.67
1 G
$499.52
5 G
$1,453.71

Vendor SKU: A903386-250MG

$237.67


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
250 MG
$237.67
1 G
$499.52
5 G
$1,453.71

About This Item

CAS Number:
Molecular Weight:
160.22
MDL number:

Vendor SKU: A903386-250MG

$237.67


Ships in 14 days - from Aldrich Partner.

Iupac Name

tert-butyl 2-ethylhydrazinecarboxylate

Inchi Code

1S/C7H16N2O2/c1-5-8-9-6(10)11-7(2,3)4/h8H,5H2,1-4H3,(H,9,10)

InChI key

AGBGVIVRTPGQSC-UHFFFAOYSA-N

storage temp.

Keep in dark place,Inert atmosphere,2-8C

Purity

97%

Country of Origin

CN

Shipping Temperature

Shipped in cold pack

Physical Form

Colorless to Yellow Solid or Semi-solid or liquid or lump

Product Link

https://www.ambeed.com/products/476362-41-1.html

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service