Skip to Content
Merck
All Photos(2)

Documents

M6773

Sigma-Aldrich

DL-Malic acid disodium salt

≥95% (capillary GC)

Synonym(s):

DL-Hydroxybutanedioic acid, DL-Hydroxysuccinic acid disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H4O5Na2
CAS Number:
Molecular Weight:
178.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95% (capillary GC)

form

powder

mp

>300 °C (lit.)

SMILES string

[Na].OC(CC(O)=O)C(O)=O

InChI

1S/C4H6O5.2Na/c5-2(4(8)9)1-3(6)7;;/h2,5H,1H2,(H,6,7)(H,8,9);;/q;2*+1/p-2

InChI key

WPUMTJGUQUYPIV-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mamoru Niikura et al.
Malaria journal, 16(1), 247-247 (2017-06-14)
Aspartate, which is converted from oxaloacetate (OAA) by aspartate aminotransferase, is considered an important precursor for purine salvage and pyrimidine de novo biosynthesis, and is thus indispensable for the growth of Plasmodium parasites at the asexual blood stages. OAA can
Shingo Sakamoto et al.
Biotechnology for biofuels, 13, 97-97 (2020-06-10)
During the chemical and biochemical decomposition of lignocellulosic biomasses, lignin is highly recalcitrant. Genetic transformation of plants to qualitatively and/or quantitatively modify lignin may reduce these recalcitrant properties. Efficient discovery of genes to achieve lignin manipulation is thus required. To
Hyuck Jun Jung et al.
Molecules (Basel, Switzerland), 23(4) (2018-03-23)
The oral bioavailability of entecavir (EV), an anti-viral agent commonly prescribed to treat hepatitis B infections, is drastically reduced under a post-prandial state. This is primarily due to its low permeability in the gastrointestinal tract. To reduce the food effect
Daniel León-Aparicio et al.
International journal of molecular sciences, 20(20) (2019-10-12)
Hyperpolarization-activated cationic HCN channels comprise four members (HCN1-4) that control dendritic integration, synaptic transmission and action potential firing. In the kidney, HCN1, HCN2 and HCN3 are differentially expressed and contribute to the transport of sodium, potassium (K+) and ammonium into
Omar Emiliano Aparicio-Trejo et al.
International journal of molecular sciences, 21(18) (2020-09-10)
Five-sixths nephrectomy (5/6Nx) model is widely used for studying the mechanisms involved in chronic kidney disease (CKD) progression, a kidney pathology that has increased dramatically in recent years. Mitochondrial impairment is a key mechanism that aggravates CKD progression; however, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service