Skip to Content
Merck
All Photos(1)

Documents

M62654

Sigma-Aldrich

3-Methyl-4-nitrophenol

98%

Synonym(s):

4-Nitro-m-cresol, 5-Hydroxy-2-nitrotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)OH
CAS Number:
Molecular Weight:
153.14
Beilstein:
1868105
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

125-130 °C (lit.)

SMILES string

Cc1cc(O)ccc1[N+]([O-])=O

InChI

1S/C7H7NO3/c1-5-4-6(9)2-3-7(5)8(10)11/h2-4,9H,1H3

InChI key

PIIZYNQECPTVEO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Galeano-Díaz et al.
Analytica chimica acta, 618(2), 131-139 (2008-06-03)
A method, using stripping square wave voltammetry (Ad-SSWV), for the simultaneous determination of fenitrothion (FEN) and its metabolites: fenitrooxon (OXON) and 3-methyl-4-nitrophenol (3-MET) in environmental samples is reported. All three compounds produce, at mercury electrode (HMDE), an electrochemical signal due
Chie Furuta et al.
Toxicology and applied pharmacology, 229(1), 109-120 (2008-03-14)
Studies of nitrophenols isolated from diesel exhaust particles (DEPs), 3-methyl-4-nitrophenol (PNMC) and 4-nitro-3-phenylphenol (PNMPP) have revealed that these chemicals possess estrogenic and anti-androgenic activity in vitro and in vivo and that PNMC accumulate in adrenal glands in vivo. However, the
S M A D Zayed et al.
Chemosphere, 70(9), 1653-1659 (2007-09-08)
The decomposition of (14)C-fenitrothion on silica gel chromatoplates as well as in polar and non polar solvents under sunlight and ultraviolet light was investigated, Its stability to sunlight on leaf surfaces of bean plants and on different surfaces (such as
Keiko Noguchi et al.
Biomedical chromatography : BMC, 21(11), 1135-1142 (2007-06-23)
Diesel exhaust particulate extract (DEPE) was obtained from diesel exhaust particulates with Soxhlet extraction using dichloromethane. After separating DEPE into 11 fractions by liquid-liquid extraction, the neutral fraction (N) showed anti-estrogenic activity and the weak acid (phenol) fraction (WA(P)) showed
Yuling Mi et al.
Toxicology letters, 190(1), 61-65 (2009-07-15)
Diesel exhaust particles (DEP) are considered to be one of the most important air pollutants. In this study, the protective effect of quercetin, an antioxidant flavonoid, on oxidative damage of testicular cells was studied by analysis of the intracellular antioxidant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service