Skip to Content
Merck
All Photos(1)

Key Documents

D31400

Supelco

Dibenz[a,h]anthracene

97%

Synonym(s):

1,2:5,6-Dibenzanthracene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14
CAS Number:
Molecular Weight:
278.35
Beilstein:
1912416
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

Assay

97%

bp

524 °C (lit.)

mp

262-265 °C (lit.)

SMILES string

c1ccc2c(c1)ccc3cc4c(ccc5ccccc45)cc23

InChI

1S/C22H14/c1-3-7-19-15(5-1)9-11-17-14-22-18(13-21(17)19)12-10-16-6-2-4-8-20(16)22/h1-14H

InChI key

LHRCREOYAASXPZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Souleymane Coulibaly et al.
Biological & pharmaceutical bulletin, 38(9), 1395-1403 (2015-09-04)
To clarify the seasonal fluctuations in air pollution and the effect of long-range transport, we collected airborne particles (n=118) at Dazaifu in Fukuoka, Japan, from June 2012 to May 2013 and measured Pb and SO4(2-), which are indicators of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service