Documents
BR23810
BRAND® compact automatic SILBERBRAND burette, without support/clamp
volume 25 mL, with LDPE bottle and PVC filling tube
Synonym(s):
Automatic Burettes, Burettes, Compact Automatic Burettes, SILBERBRAND
About This Item
UNSPSC Code:
41121808
NACRES:
NB.43
material
AR-Glass
manufacturer/tradename
BRAND 23810
volume measuring range
25 mL, accuracy: ±0.08 mL
volume
25 mL
General description
The Modular Automatic Burette Concept! Fast to dismantle and easy to clean. Easy to repair - all individual components replaceable! Boro 3.3, detachable PTFE stopcock. Error limits according to class B, DIN EN ISO 385. Calibrated to deliver (TD, Ex).
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG