B2608
1,9-Benz-10-anthrone
technical grade
Synonym(s):
7H-Benz[de]anthracen-7-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C17H10O
CAS Number:
Molecular Weight:
230.26
Beilstein:
1455646
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
technical grade
mp
168-170 °C (lit.)
SMILES string
O=C1c2ccccc2-c3cccc4cccc1c34
Looking for similar products? Visit Product Comparison Guide
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service