Skip to Content
Merck
All Photos(1)

Key Documents

86615

Sigma-Aldrich

Tetraethylammonium fluoride dihydrate

≥97.0% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5)4N(F) · 2H2O
CAS Number:
Molecular Weight:
185.28
Beilstein:
4152659
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥97.0% (T)

storage temp.

2-8°C

SMILES string

O.O.[F-].CC[N+](CC)(CC)CC

InChI

1S/C8H20N.FH.2H2O/c1-5-9(6-2,7-3)8-4;;;/h5-8H2,1-4H3;1H;2*1H2/q+1;;;/p-1

InChI key

HRTKQUHFGZFPPF-UHFFFAOYSA-M

Application

  • Catalyst for oxidation, methylation, and deuteration reactions of fluorene

Other Notes

Reagent for the cleavage of Si-C bonds.; Review of its use as a base in organic synthesis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.H. Clark
Chemical Reviews, 80, 429-429 (1980)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service