Skip to Content
Merck
All Photos(2)

Key Documents

795232

Sigma-Aldrich

1-Bromovinyl-1-boronic acid MIDA ester

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H9BBrNO4
CAS Number:
Molecular Weight:
261.87
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

153-158 °C

SMILES string

C=C(Br)B1OC(CN(C)CC(O1)=O)=O

InChI

1S/C7H9BBrNO4/c1-5(9)8-13-6(11)3-10(2)4-7(12)14-8/h1,3-4H2,2H3

InChI key

PZDXKEHNRTYTFO-UHFFFAOYSA-N

Application

This MIDA Boronate building block has been used in highly chemoselective sequences toward the synthesis of 1; 1-disubstituted olefins.

(1-Bromovinyl)-MIDA boronate: a readily accessible and highly versatile building block for small molecule synthesis

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service