Skip to Content
Merck
All Photos(1)

Documents

721395

Sigma-Aldrich

Ammonium hydroxide-18O solution

~3 N in H218O, 95 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH418OH/H218O
Molecular Weight:
37.05
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

isotopic purity

95 atom % 18O

form

solution

concentration

~3 N in H218O

mass shift

M+2

storage temp.

2-8°C

SMILES string

[NH4+].[18OH-]

InChI

1S/H3N.H2O/h1H3;1H2/i;1+2

InChI key

VHUUQVKOLVNVRT-NLQOEHMXSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service