Skip to Content
Merck
All Photos(1)

Documents

71585

Sigma-Aldrich

4-Chloro-3-fluoropyridine

≥95.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H3ClFN
CAS Number:
Molecular Weight:
131.54
Beilstein:
1422747
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0%

form

liquid

storage condition

under inert gas (Argon)

density

1.333 g/mL at 25 °C (lit.)

shipped in

wet ice

storage temp.

2-8°C

SMILES string

Fc1cnccc1Cl

InChI

1S/C5H3ClFN/c6-4-1-2-8-3-5(4)7/h1-3H

InChI key

BEQUUSCRAKEKQM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

may discolor to yellow on storage

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

82.4 °F - closed cup

Flash Point(C)

28 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service