Skip to Content
Merck
All Photos(1)

Documents

714976

Sigma-Aldrich

Bis(2,4-dimethylpentadienyl)ruthenium(II)

99%

Synonym(s):

Bis(η5-2,4-dimethylpentadienyl)ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ru(C7H11)2
CAS Number:
Molecular Weight:
291.39
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

powder, crystals or beads

reaction suitability

core: ruthenium
reagent type: catalyst

mp

91-93 °C

SMILES string

[Ru].[CH2][C](C)[CH][C]([CH2])C.[CH2][C](C)[CH][C]([CH2])C

InChI

1S/2C7H11.Ru/c2*1-6(2)5-7(3)4;/h2*5H,1,3H2,2,4H3;

InChI key

ZOLKMBFKHJHDCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service