Skip to Content
Merck
All Photos(2)

Documents

695491

Sigma-Aldrich

(1R,3S,4S)-N-Boc-2-azabicyclo[2.2.1]heptane-3-carboxylic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19NO4
CAS Number:
Molecular Weight:
241.28
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

powder

optical activity

[α]22/D -170±10°, c = 1 in chloroform

mp

147-152 °C

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N1C2CCC(C2)[C@H]1C(O)=O

InChI

1S/C12H19NO4/c1-12(2,3)17-11(16)13-8-5-4-7(6-8)9(13)10(14)15/h7-9H,4-6H2,1-3H3,(H,14,15)/t7-,8+,9-/m0/s1

InChI key

IFAMSTPTNRJBRG-YIZRAAEISA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 36, 6765-6765 (1995)
Srikanth Venkatraman et al.
Bioorganic & medicinal chemistry letters, 16(6), 1628-1632 (2006-01-18)
Prolonged hepatitis C infection is the leading cause for cirrhosis of the liver and hepatocellular carcinoma. The etiological agent HCV virus codes a single polyprotein of approximately 3000 amino acids that is processed with the help of a serine protease

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service