Skip to Content
Merck
All Photos(1)

Documents

671738

Sigma-Aldrich

1,3-Dimethylimidazolium methanesulfonate

for catalysis, ≥99.0% (HPLC/T)

Synonym(s):

MMIM CH3SO3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12N2O3S
CAS Number:
Molecular Weight:
192.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

for catalysis

Assay

≥99.0% (HPLC/T)

impurities

≤10 mg/kg halogenes
≤200 ppm water

SMILES string

CS([O-])(=O)=O.Cn1cc[n+](C)c1

InChI

1S/C5H9N2.CH4O3S/c1-6-3-4-7(2)5-6;1-5(2,3)4/h3-5H,1-2H3;1H3,(H,2,3,4)/q+1;/p-1

InChI key

CUCZSYMTPDSMEL-UHFFFAOYSA-M

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiheng Chu et al.
Environmental science & technology, 49(9), 5511-5519 (2015-04-02)
Photochemical transformations greatly affect the stability and fate of amino acids (AAs) in sunlit aquatic ecosystems. Whereas the direct phototransformation of dissolved AAs is well investigated, their indirect photolysis in the presence of chromophoric dissolved organic matter (CDOM) is poorly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service