Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

616923

Methyl-d3-triphenylphosphonium iodide

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3P(C6H5)3I
CAS Number:
Molecular Weight:
407.24
MDL number:

Pricing and availability is not currently available.

isotopic purity

98 atom % D

mp

186-188 °C (lit.)

mass shift

M+3

SMILES string

[I-].[2H]C([2H])([2H])[P+](c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C19H18P.HI/c1-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19;/h2-16H,1H3;1H/q+1;/p-1/i1D3;

InChI key

JNMIXMFEVJHFNY-NIIDSAIPSA-M

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service