Skip to Content
Merck
All Photos(1)

Documents

568929

Sigma-Aldrich

Tolylene-2,4-diisothiocyanate

97%

Synonym(s):

2,4-Toluene diisothiocyanate, 4-Methyl-1,3-phenylene diisothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NCS)2
CAS Number:
Molecular Weight:
206.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

54-57 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1ccc(cc1N=C=S)N=C=S

InChI

1S/C9H6N2S2/c1-7-2-3-8(10-5-12)4-9(7)11-6-13/h2-4H,1H3

InChI key

NNMVCFPMIBOZCL-UHFFFAOYSA-N

Caution

Refrigerate

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service