Skip to Content
Merck
All Photos(1)

Documents

463094

Sigma-Aldrich

Trimethylboron

electronic grade, ≥98.35%

Synonym(s):

Trimethylborane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
B(CH3)3
CAS Number:
Molecular Weight:
55.91
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

electronic grade

Assay

≥98.35%

form

gas

reaction suitability

core: boron

impurities

<1 ppm Oxygen (O2)
<1.5% Dimethylethylborane [B(CH3)2C2H5]
<10 ppm Carbon dioxide (CO2)
<10 ppm Nitrogen (N2)
<100 ppm Methane (CH4)
<1000 ppm Trimethylamine (TMA)
<50 ppm Hydrogen (H2)

bp

−20.2 °C (lit.)

mp

−161.5 °C (lit.)

SMILES string

CB(C)C

InChI

1S/C3H9B/c1-4(2)3/h1-3H3

InChI key

WXRGABKACDFXMG-UHFFFAOYSA-N

Application

Precursor to boron carbide thin films by chemical vapor deposition (CVD).
Trimethylboron is used as an organometallic precursor for the CVD of boron-carbon containing films and as a boron source in the deposition of borophosphosilicate glass (BPSG).

Recommended products

Stainless steel regulators Z527416 or Z527424 are recommended.

Pictograms

Gas cylinderCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Press. Gas Liquefied gas - Skin Corr. 1B

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Longxin Chen et al.
Science advances, 6(14), eaaz7825-eaaz7825 (2020-04-10)
Currently, there are no methods available offering solutions to select and identify antibodies binding to a specific conformational epitope of an antigen. Here, we developed a method to allow epitope-directed antibody selection from a phage display library by photocrosslinking bound
Materials Letters, 27, 327-327 (1996)
Balázs Koscsó et al.
Science immunology, 5(46) (2020-04-12)
Intestinal mononuclear phagocytes (MPs) are composed of heterogeneous dendritic cell (DC) and macrophage subsets necessary for the initiation of immune response and control of inflammation. Although MPs in the normal intestine have been extensively studied, the heterogeneity and function of
Maha Almanan et al.
Science advances, 6(31), eabb0806-eabb0806 (2020-08-25)
Aging results in profound immune dysfunction, resulting in the decline of vaccine responsiveness previously attributed to irreversible defects in the immune system. In addition to increased interleukin-6 (IL-6), we found aged mice exhibit increased systemic IL-10 that requires forkhead box
Emma K Persson et al.
Science (New York, N.Y.), 364(6442) (2019-05-28)
Although spontaneous protein crystallization is a rare event in vivo, Charcot-Leyden crystals (CLCs) consisting of galectin-10 (Gal10) protein are frequently observed in eosinophilic diseases, such as asthma. We found that CLCs derived from patients showed crystal packing and Gal10 structure

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service