Skip to Content
Merck
All Photos(2)

Key Documents

447471

Sigma-Aldrich

Methyl 10-hydroxydecanoate

technical grade, ≥90%

Synonym(s):

Pd/C

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$249.00

$249.00


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
10 G
$249.00

About This Item

Linear Formula:
HO(CH2)9CO2CH3
CAS Number:
Molecular Weight:
202.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$249.00


Estimated to ship on23 April 2025


Request a Bulk Order

grade

technical grade

Assay

≥90%

refractive index

n20/D 1.448 (lit.)

density

0.962 g/mL at 25 °C (lit.)

functional group

ester
hydroxyl

SMILES string

COC(=O)CCCCCCCCCO

InChI

1S/C11H22O3/c1-14-11(13)9-7-5-3-2-4-6-8-10-12/h12H,2-10H2,1H3

InChI key

NFJQGHRDVMDLKM-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

221.0 °F - closed cup

Flash Point(C)

105.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Notes-A Preparation of 10-Hydroxydecanoic Acid.
Benton F and Kiess F.
The Journal of Organic Chemistry, 25(3), 470-471 (1960)
N M Carballeira et al.
Chemistry and physics of lipids, 97(2), 87-91 (1999-04-08)
The marine bacterial fatty acid 9-methyl-10-hexadecenoic acid was conveniently prepared in 6 steps and in a 22% overall yield, starting from commercially available methyl 10-hydroxydecanoate. The naturally occurring fatty acid has the E double bond configuration as confirmed by gas
Synthesis and polymerization of carboxyalkylmethacrylates: new hydrophobic polyelectrolytes.
Rogel-Hernandez E, et al.
Designed Monomers and Polymers, 4(4), 343-356 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service