Skip to Content
Merck
All Photos(2)

Key Documents

432199

Sigma-Aldrich

Leucoberbelin Blue I

Dye content 65 %

Synonym(s):

LBB

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$91.50
500 MG
$319.00

$91.50


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$91.50
500 MG
$319.00

About This Item

Empirical Formula (Hill Notation):
C23H26N2O3S
CAS Number:
Molecular Weight:
410.53
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

$91.50


Estimated to ship on26 April 2025


Request a Bulk Order

form

powder

composition

Dye content, 65%

technique(s)

microbe id | staining: suitable

mp

>300 °C (lit.)

solubility

water: 50 g/L

λmax

253 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

CN(C)c1ccc(cc1)C(c2ccc(cc2)N(C)C)c3ccccc3S(O)(=O)=O

InChI

1S/C23H26N2O3S/c1-24(2)19-13-9-17(10-14-19)23(18-11-15-20(16-12-18)25(3)4)21-7-5-6-8-22(21)29(26,27)28/h5-16,23H,1-4H3,(H,26,27,28)

InChI key

MCKLFIWDQVFMEK-UHFFFAOYSA-N

General description

Leucoberbelin Blue I is a colorimetric reagent redox dye. It is a water-soluble leuco base also abbreviated as LBB. Upon oxidation, LBB forms a colored complex which is stoichiometrically related to the oxidation state of the reagent.

Application

Leucoberbelin blue I has been used as a quencher in the time-resolved fluorescence QRET technique for receptor-ligand interaction screening. LBB assay is used to indicate the kinetics of Mn (II) oxides formation.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natália R Barboza et al.
Frontiers in microbiology, 8, 1946-1946 (2017-10-25)
Manganese is an important metal for the maintenance of several biological functions, but it can be toxic in high concentrations. One of the main forms of human exposure to metals, such as manganese (Mn), is the consumption of solar salt
Monica Agnolucci et al.
Frontiers in plant science, 9, 1956-1956 (2019-01-30)
The implementation of sustainable agriculture encompasses practices enhancing the activity of beneficial soil microorganisms, able to modulate biogeochemical soil cycles and to affect soil fertility. Among them, arbuscular mycorrhizal fungi (AMF) establish symbioses with the roots of most food crops
Deric R Learman et al.
Frontiers in microbiology, 4, 262-262 (2013-09-13)
Manganese (Mn) oxides are among the most reactive sorbents and oxidants within the environment, where they play a central role in the cycling of nutrients, metals, and carbon. Recent discoveries have identified superoxide ([Formula: see text]) both of biogenic and
Giulia Castorina et al.
Plant physiology, 184(1), 266-282 (2020-07-16)
In all land plants, the outer surface of aerial parts is covered by the cuticle, a complex lipid layer that constitutes a barrier against damage caused by environmental factors and provides protection against nonstomatal water loss. We show in this
E R Estes et al.
Geobiology, 15(1), 158-172 (2016-07-12)
Manganese (Mn) oxides participate in a range of interactions with organic carbon (OC) that can lead to either carbon degradation or preservation. Here, we examine the abundance and composition of OC associated with biogenic and environmental Mn oxides to elucidate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service