Skip to Content
Merck
All Photos(1)

Key Documents

419583

Sigma-Aldrich

2-Butenylmagnesium chloride solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH2MgCl
CAS Number:
Molecular Weight:
114.86
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

density

0.921 g/mL at 25 °C

SMILES string

C\C=C\C[Mg]Cl

InChI

1S/C4H7.ClH.Mg/c1-3-4-2;;/h3-4H,1H2,2H3;1H;/q;;+1/p-1/b4-3+;;

InChI key

RTJGBPYEBKZQGE-CZEFNJPISA-M

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F

Flash Point(C)

-17 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service