Skip to Content
Merck
All Photos(3)

Documents

389633

Sigma-Aldrich

Bromochloroacetic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrClCHCOOH
CAS Number:
Molecular Weight:
173.39
Beilstein:
1720556
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.51 (lit.)

bp

210-212 °C/767 mmHg (lit.)

mp

27.5 °C (lit.)

density

1.985 g/mL at 25 °C (lit.)

SMILES string

[H]C(Cl)(Br)C(O)=O

InChI

1S/C2H2BrClO2/c3-1(4)2(5)6/h1H,(H,5,6)

InChI key

GEHJBWKLJVFKPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Douglas B Tully et al.
Reproductive toxicology (Elmsford, N.Y.), 19(3), 353-366 (2005-02-03)
A byproduct of drinking water disinfection, bromochloroacetic acid (BCA), acts as a reproductive toxicant in rats. To determine if BCA produces similar reproductive toxicity in mice, juvenile and adult C57BL/6 males were exposed to 0, 8, 24, 72 or 216
B Ells et al.
Analytical chemistry, 71(20), 4747-4752 (1999-11-05)
The lower limit of detection for low molecular weight polar and ionic analytes using electrospray ionization-mass spectrometry (ESI-MS) is often severely compromised by an intense background that obscures ions of trace components in solution. Recently, a new technique, referred to
D Martínez et al.
Journal of chromatography. A, 808(1-2), 229-236 (1998-07-04)
A capillary zone electrophoresis (CZE) system for determining haloacetic acids in water was optimized with indirect photometric detection. Two different carrier electrolytes, potassium hydrogenphthalate and sodium 2,6-naphthalenedicarboxylate, were evaluated in terms of sensitivity and two different electroosmotic flow modifiers, tetradecyltrimethylammonium
J M Parrish et al.
Toxicology, 110(1-3), 103-111 (1996-06-17)
Brominated and chlorinated haloacetates (HAs) are by-products of drinking water disinfection. Dichloroacetate (DCA) and trichloroacetate (TCA) are hepatocarcinogenic in rodents, but the brominated analogs have received little study. Prior work has indicated that acute doses of the brominated derivatives are
Xiaoling Zhang et al.
Toxicology, 289(2-3), 151-159 (2011-08-26)
Bromate (BrO(3)(-)) is a drinking water disinfection by-product (DBP) that induces renal cell death via DNA damage-dependent and -independent mechanisms. Drinking water contains other DBPs in addition to BrO(3)(-). We tested the effect of two of these, sodium chlorite (NaClO(2))

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service