Skip to Content
Merck
All Photos(1)

Key Documents

378593

Supelco

IWT® TMD-8 hydrogen and hydroxide form

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12000000

description

self-indicating mixed bed resin

parameter

41 °C max. temp.

matrix

gel

particle size

16-40 mesh

operating pH

0-14

capacity

0.55 meq/mL by wetted bed volume
0.80 meq/g by dry weight

General description

A mixture of strong cation and strong anion exchange resins, with capacity indicator. Selective beads change color from blue-green to amber when exchange capacity is reached.

Application

Self-indicating resin. Changes color from blue-green to amber when exchange capacity reached

Legal Information

IWT is a registered trademark of U.S. Filter Corp.

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service