Skip to Content
Merck
All Photos(2)

Documents

308145

Sigma-Aldrich

N-Propylethylenediamine

≥97%

Synonym(s):

2-Propylaminoethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2NHCH2CH2NH2
CAS Number:
Molecular Weight:
102.18
Beilstein:
1697232
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97%

form

liquid

refractive index

n20/D 1.441 (lit.)

bp

147-150 °C (lit.)

density

0.829 g/mL at 25 °C (lit.)

SMILES string

CCCNCCN

InChI

1S/C5H14N2/c1-2-4-7-5-3-6/h7H,2-6H2,1H3

InChI key

CFNHVUGPXZUTRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

N-Propylethylenediamine columns help in quantitatively removing Cu(2+) from buffered solutions at pH 6.

Application

N-Propylethylenediamine was used in the synthesis of N-acetyl-N′-propylethylenediamine.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F - closed cup

Flash Point(C)

45 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Murakami Y, et al.
Tetrahedron Letters, 38(21), 3751-3754 (1997)
Yanping Li et al.
Journal of separation science, 43(8), 1466-1473 (2020-02-14)
A method for determining amitraz and 2,4-dimethylaniline in honey was established by using ultra-high-performance liquid chromatoghaphy and Q Exactive after applying quick, easy, cheap, effective, rugged, and safe extracting process. A suitable extraction method was designed to extract the amitraz and
Xiaohong Yang et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 55(5), 477-483 (2020-05-26)
Pyraoxystrobin is a novel strobilurin fungicide that is widely used on many crops. The high log Kow of pyraoxystrobin implies that it tends to accumulate in aquatic organisms. This study optimized the sorbents of QuEChERS (quick, easy, cheap, effective, rugged
D K Ryan et al.
Talanta, 32(9), 859-863 (1985-09-01)
Three types of chelating agents immobilized on glass were compared with Chelex 100 for removing and preconcentrating trace Cu(2+) from laboratory-prepared solutions. Columns of immobilized N-propylethylenediamine (diamine), its bis(dithiocarbamate) (DTC) and immobilized 8-hydroxyquinoline (8-HQ) quantitatively remove Cu(2+) (10-200 mug l.
Xu Yu et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-08)
Rapeseed (Brassica napus L.) is rich in phenols, vitamins, carotenoids, and mineral elements, such as selenium. Additionally, it contains the active ingredients sulforaphane and indole-3-carbinol, which have been demonstrated to have pharmacological effects. In this study, sulforaphane and indole-3-carbinol were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service