Skip to Content
Merck
All Photos(1)

Documents

301396

Sigma-Aldrich

4-(Trifluoromethoxy)benzoic acid

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4CO2H
CAS Number:
Molecular Weight:
206.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

150-154 °C (lit.)

SMILES string

OC(=O)c1ccc(OC(F)(F)F)cc1

InChI

1S/C8H5F3O3/c9-8(10,11)14-6-3-1-5(2-4-6)7(12)13/h1-4H,(H,12,13)

InChI key

RATSANVPHHXDCT-UHFFFAOYSA-N

General description

Kinetics of internal acyl migration and hydrolysis of the synthetic β-1-O-acyl-D-glucopyranuronates of 2-, 3-, and 4-(trifluoromethyl)benzoic acids in phosphate buffer was studied.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A W Nicholls et al.
Chemical research in toxicology, 9(8), 1414-1424 (1996-12-01)
High resolution 19F NMR spectroscopy has been used to investigate the kinetics of internal acyl migration and hydrolysis of the synthetic beta -1-O-acyl-D-glucopyranuronates of 2-, 3-, and 4-(trifluoromethyl) benzoic acids (TFMBAs) in phosphate buffer solutions at 30 degrees C as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service