Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

284866

Sigma-Aldrich

2-Nitro-5-thiocyanatobenzoic acid

98%

Synonym(s):

2-Nitro-5-thiocyanobenzoic acid, NTCB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(SCN)CO2H
CAS Number:
Molecular Weight:
224.19
Beilstein:
2124001
EC Number:
MDL number:
UNSPSC Code:
12352103

Pricing and availability is not currently available. Contact Technical Service

Assay

98%

mp

156-157 °C (lit.)

SMILES string

OC(=O)c1cc(SC#N)ccc1[N+]([O-])=O

InChI

1S/C8H4N2O4S/c9-4-15-5-1-2-7(10(13)14)6(3-5)8(11)12/h1-3H,(H,11,12)

InChI key

NQUNIMFHIWQQGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service