Skip to Content
Merck
All Photos(1)

Documents

281891

Sigma-Aldrich

1-Phenylpiperazine hydrochloride

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2 · HCl
CAS Number:
Molecular Weight:
198.69
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

247-250 °C (lit.)

SMILES string

Cl[H].C1CN(CCN1)c2ccccc2

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cemal Parlak et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 82(1), 383-388 (2011-08-09)
New Hofmann type clathrates in the form of M(pp)(2)Ni(CN)(4)·2G (where pp = 1-phenylpiperazine, G = 1,4-dioxane and M = Ni, Co or Cd) have been prepared in powder form and their FT-IR and FT-Raman spectra are reported. The results suggest
Jacek Sapa et al.
Pharmacological reports : PR, 63(1), 71-78 (2011-03-29)
The present study was designed to investigate the central nervous system activity of 23 novel phenylpiperazine pyrrolidin-2-one derivatives. These compounds had marked antiarrhythmic and hypotensive activities and revealed affinity for α1- and α2-adrenoceptors. These effects may be related to their
Jadwiga Handzlik et al.
Bioorganic & medicinal chemistry, 20(7), 2290-2303 (2012-03-03)
An association between α(1)-adrenoceptor affinities, hERG K(+)-antagonistic properties and antiarrhythmic activities for a series of phenylpiperazine derivatives of hydantoin (2a-21a) was investigated. New compounds were synthesized and tested for their affinity for α(1)-adrenoceptors in radioligand binding assay using [(3)H]-prazosin as
Fabrizio Sanna et al.
Bioorganic & medicinal chemistry, 21(7), 1680-1684 (2013-02-23)
Employing the D4 selective phenylpiperazine 2 as a lead compound, planar chiral analogs with paracyclophane substructure were synthesized and evaluated for their ability to bind and activate dopamine receptors. The study revealed that the introduction of a [2.2]paracyclophane moiety is
Rodolfo do Couto Maia et al.
Expert opinion on therapeutic patents, 22(10), 1169-1178 (2012-09-11)
The N-phenylpiperazine subunit represents one of the most versatile scaffolds used in the medicinal chemistry field. Recently, some N-phenylpiperazine derivatives have reached late stage clinical trials for the treatment of CNS disorders, thus, this is clearly a molecular template that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service