Skip to Content
Merck
All Photos(1)

Key Documents

256447

Sigma-Aldrich

Bis(cyclopentadienyl)molybdenum(IV) dichloride

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10Cl2Mo
CAS Number:
Molecular Weight:
297.03
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

crystalline powder
powder, crystals or chunks

reaction suitability

core: molybdenum
reagent type: catalyst

SMILES string

Cl[Mo]Cl.[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.2ClH.Mo/c2*1-2-4-5-3-1;;;/h2*1-5H;2*1H;/q;;;;+2/p-2

InChI key

YQZUTDPJTAZPQJ-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Köpf-Maier et al.
Zeitschrift fur Naturforschung. Section C, Biosciences, 34(12), 1174-1176 (1979-12-01)
The antitumor activity of molybdocene dichloride is tested against Ehrlich ascites tumor in CF1 mice. The application of 75 of 100 mg/kg 24 h after transplantation achieves 100% tumor inhibition until day 30. Following the d0 and d1 metallocene dichlorides
Andrea Erxleben
Inorganic chemistry, 44(4), 1082-1094 (2005-04-30)
Reactions of the organometallic compound molybdocene dichloride (Cp2MoCl2, Cp = eta5-cyclopentadienyl) with the cysteine-containing peptides L-cysteinylglycine (Cys-Gly), N-acetyl-L-cysteine (AcCys), glycyl-L-cysteine (Gly-Cys), glycyl-L-cysteinylglycine (Gly-Cys-Gly), and gamma-L-glutamyl-L-cysteinylglycine (glutathione, GSH) have been studied in aqueous solution in the pH range 2-9. The dipeptides
Jenny B Waern et al.
Inorganic chemistry, 43(1), 206-213 (2004-01-06)
The relative affinity of molybdocene dichloride (Cp(2)MoCl(2)) for the thiol, amino, carboxylate, phosphate(O) and heterocyclic(N) donor ligands present in amino acids and nucleotides, has been studied in aqueous solutions at pH 2-7, using (1)H, (13)C and (31)P NMR spectroscopy. Molybdocene
E Tonew et al.
Zentralblatt fur Bakteriologie, Mikrobiologie und Hygiene. 1. Abt. Originale A, Medizinische Mikrobiologie, Infektionskrankheiten und Parasitologie = International journal of microbiology and hygiene. A, Medical microbiology, infectious..., 250(4), 425-430 (1981-11-01)
In view of the fact that bis cyclopentadienyl metal dihalides are known to be anti-tumour drugs, we have investigated the antiviral activity of this type of coordination compounds. Bis cyclopentadienyl titanium dichloride (a) has shown significant antiviral efficiency in vitro
Louis Y Kuo et al.
Inorganic chemistry, 44(15), 5537-5541 (2005-07-19)
Thiophosphinate hydrolysis involving P-S bond scission is desirable for the degradation of organophosphate neurotoxins, and we report the first case for such a hydrolytic process by an organometallic compound. The metallocene, bis(cyclopentadienyl)molybdenum(IV) dichloride, Cp(2)MoCl(2) (Cp = eta(5)-C(5)H(5)), hydrolyzes a variety

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service