Skip to Content
Merck
All Photos(1)

Documents

24910

Sigma-Aldrich

1-Chloro-2-iodobenzene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4I
CAS Number:
Molecular Weight:
238.45
Beilstein:
1905116
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.6334 (lit.)
n20/D 1.635

bp

234-235 °C (lit.)

mp

1 °C (lit.)

density

1.952 g/mL at 25 °C (lit.)

SMILES string

Clc1ccccc1I

InChI

1S/C6H4ClI/c7-5-3-1-2-4-6(5)8/h1-4H

InChI key

MPEOPBCQHNWNFB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Chloro-2-iodobenzene reacts with lanthanum metal in the presence of trimethylchlorosilane to yield 1-chloro-2-trimethylsilylbenzene.

Application

1-Chloro-2-iodobenzene has been used in the preparation of N,N-diaryl-o-phenylenediamines and 4(5)-(2-chlorophenyl)-1H-imidazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pd catalyzed coupling of 1, 2-dibromoarenes and anilines: formation of N,N-diaryl-o-phenylenediamines.
Wenderski T, et al.
Tetrahedron Letters, 45(37), 6851-6853 (2004)
A new boronic-acid based strategy to synthesize 4 (5)-(het) aryl-1H-imidazoles.
Primas N, et al.
Tetrahedron, 64(20), 4596-4601 (2008)
Reaction of 1, 2-dihalogen substituted arenes with lanthanum metal: a new generation method of benzyne.
Kawabata H, et al.
Tetrahedron Letters, 43(28), 4911-4913 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service